missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Phosphate Sodium Salt, TRC

Product Code. 30138295
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138295 100 mg 100mg
30136024 1 g 1g
30005664 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138295 Supplier Toronto Research Chemicals Supplier No. TRCD477600100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32586-82-6
Molecular Formula C2 H6 O4 P . Na
Molecular Weight (g/mol) 148.03
Synonym Phosphoric acid, dimethyl ester, sodium salt (1:1) (ACI), Phosphoric acid, dimethyl ester, sodium salt (8CI, 9CI), Dimethyl monosodium phosphate, Dimethyl sodium phosphate, Methyl sodium phosphite ((MeO)2(NaO)PO), Monosodium dimethyl phosphate, Sodium dimethyl phosphate, Dimethyl phosphate sodium salt
IUPAC Name sodium;dimethyl phosphate
SMILES [Na+].COP(=O)([O-])OC

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C2H7O4P.Na/c1-5-7(3,4)6-2;/h1-2H3,(H,3,4);/q;+1/p-1
Formula Weight 147.9901
Chemical Name or Material Dimethyl Phosphate Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.