Learn More
Dimethyl biphenyl-4,4'-dicarboxylate, 98%, Thermo Scientific™
Product Code. p-7020559
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424254 1 g 1g
11434254 5 g 5g
11444254 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11424254 Supplier Thermo Scientific Alfa Aesar Supplier No. A12556.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Dimethyl biphenyl-4,4′-dicarboxylate is used as a hepatoprotectant in the treatment of chronic hepatitis. At 250 µg/ml, it has been shown to stimulate JAK/STAT signaling and to induce the expression of interferon-α stimulated genes in a HepG2 cell line. It has been used to study hepatic fibrosis and chronic hepatocellular damage. It appears to normalize alanine aminotransferase (ALT) in cases where levels of ALT are elevated. It may be employed as starting reagent for the synthesis of dimethyl 2-fluoro-and 2,2′-difluorobiphenyl-4,4′-dicarboxylates.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 792-74-5
Molecular Formula C16H14O4
Molecular Weight (g/mol) 270.28
MDL Number MFCD00017201
InChI Key BKRIRZXWWALTPU-UHFFFAOYSA-N
Synonym dimethyl 1,1'-biphenyl-4,4'-dicarboxylate, dimethyl biphenyl-4,4'-dicarboxylate, biphenyl dimethyl dicarboxylate, dimethyl 4,4'-biphenyldicarboxylate, 1,1'-biphenyl-4,4'-dicarboxylic acid, dimethyl ester, unii-k61bxa0u9c, 4,4'-biphenyldicarboxylic acid, dimethyl ester, k61bxa0u9c, methyl 4-4-methoxycarbonylphenyl benzoate, methyl 4-4-methoxycarbonyl phenyl benzoate
PubChem CID 13099
IUPAC Name methyl 4-(4-methoxycarbonylphenyl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)C1=CC=C(C=C1)C(=O)OC
Specifications
Melting Point 212°C to 216°C
Quantity 1 g
Beilstein 2055852
Solubility Information Insoluble in water.
Formula Weight 270.29
Percent Purity 98%
Chemical Name or Material Dimethyl biphenyl-4,4'-dicarboxylate