missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (4-methoxybenzyl)malonate, 95%, Thermo Scientific™

Product Code. 11401220
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401220 1 g 1g
11411220 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11401220 Supplier Thermo Scientific Alfa Aesar Supplier No. H26331.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15378-09-3
Molecular Formula C13H16O5
Molecular Weight (g/mol) 252.266
MDL Number MFCD08436113
InChI Key GXDFOJRQIDXKIH-UHFFFAOYSA-N
Synonym dimethyl 2-4-methoxybenzyl malonate, 4-methoxybenzyl malonic acid dimethyl ester, 1,3-dimethyl 2-4-methoxyphenyl methyl propanedioate, acmc-1bt9b, dimethyl 4-methoxybenzyl malonate, dimethyl 2 4-methoxybenzyl malonate, dimethyl 4-methoxyphenyl methyl propanedioate, propanedioic acid,2-4-methoxyphenyl methyl-, 1,3-dimethyl ester
PubChem CID 12567675
IUPAC Name dimethyl 2-[(4-methoxyphenyl)methyl]propanedioate
SMILES COC1=CC=C(C=C1)CC(C(=O)OC)C(=O)OC

Specifications

Quantity 1 g
Formula Weight 252.27
Percent Purity 95%
Chemical Name or Material Dimethyl (4-methoxybenzyl)malonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.