missing translation for 'onlineSavingsMsg'
Learn More

Dimethicone ~2000 (Polydimethylsiloxane), TRC

Product Code. 30136402
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136402 100 g 100g
30132735 25 g 25g
30005263 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136402 Supplier Toronto Research Chemicals Supplier No. TRCD461510100G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9016-00-6
Molecular Formula C3H9Si(C2H6OSi)nCH3
Molecular Weight (g/mol) 74.15
Synonym Di-Me Polysiloxane, Di-Me Silicone, Di-Me Siloxane, Di-Me Siloxane, SRU, Dimethicone 200SF, Dimethyl Siloxane, Dimethyl Siloxane, sru, Dimethyldichlorosilane Hydrolytic Polymer, sru, Dimethylethoxysilane Homopolymer, sru, Dimethylpolysiloxane, Dimethylpolysiloxanes, Dimethylsilanediol Homopolymer, SRU, Dimethylsilanediol Polymer, sru, Dimethylsilicone, Dimethylsiloxane Homopolymer, SRU
IUPAC Name 1,1,1,3,3,3-hexamethyldisiloxane polymer
SMILES C[Si](C)(C)O[Si](C)(C)C

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H18OSi2/c1-8(2,3)7-9(4,5)6/h1-6H3
Formula Weight 74.0188
Chemical Name or Material Polydimethylsiloxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.