missing translation for 'onlineSavingsMsg'
Learn More

Dimethenamide-P, TRC

Product Code. 30137471
Change view
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137471 50 mg 50mg
17942835 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137471 Supplier Toronto Research Chemicals Supplier No. TRCD46058350MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163515-14-8
Molecular Formula C12 H18 Cl N O2 S
Molecular Weight (g/mol) 275.79
Synonym Acetamide, 2-chloro-N-(2,4-dimethyl-3-thienyl)-N-(2-methoxy-1-methylethyl)-, (S)-, 2-Chloro-N-(2,4-dimethyl-3-thienyl)-N-[(1S)-2-methoxy-1-methylethyl]acetamide, (S)-Dimethenamid, BAS 656H, DMTA-P, Dimethenamid P, Establish, Frontier X 2, Isard, Outlook, Spectrum, Spectrum (herbicide), Tower (herbicide)
IUPAC Name 2-chloro-N-(2,4-dimethylthiophen-3-yl)-N-[(2S)-1-methoxypropan-2-yl]acetamide
SMILES COC[C@H](C)N(C(=O)CCl)c1c(C)csc1C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H18ClNO2S/c1-8-7-17-10(3)12(8)14(11(15)5-13)9(2)6-16-4/h7,9H,5-6H2,1-4H3/t9-/m0/s1
Formula Weight 275.0747
Chemical Name or Material Dimethenamid-P
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.