Learn More
Dimethenamid, TRC
Product Code. 17938159
Change view
Click to view available options
Quantity:
5 mg
10 mg
100 mg
Unit Size:
100mg
10mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17938159 5 mg 5mg
17998879 100 mg 100mg
17948899 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17938159 Supplier Toronto Research Chemicals Supplier No. TRCD4605805MG
 Restricted Item
Estimated Shipment: 16-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 87674-68-8
Molecular Formula C12 H18 Cl N O2 S
Molecular Weight (g/mol) 275.79
Synonym 2-Chloro-N-(2,4-dimethyl-3-thienyl)-N-(2-methoxy-1-methylethyl)acetamide, Dimethenamid, Fieldstar, Frontier, SAN 582, SAN 582H
IUPAC Name 2-chloro-N-(2,4-dimethylthiophen-3-yl)-N-(1-methoxypropan-2-yl)acetamide
SMILES COCC(C)N(C(=O)CCl)c1c(C)csc1C
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C12H18ClNO2S/c1-8-7-17-10(3)12(8)14(11(15)5-13)9(2)6-16-4/h7,9H,5-6H2,1-4H3
Formula Weight 275.0747
Chemical Name or Material Dimethenamid