missing translation for 'onlineSavingsMsg'
Learn More

Dimepiperate, TRC

Product Code. 30381417
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381417 5 mg
30371763 10 mg
30379619 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381417 Supplier Toronto Research Chemicals Supplier No. TRCD4602505MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61432-55-1
Molecular Formula C15 H21 N O S
Molecular Weight (g/mol) 263.4
Synonym Dimepiperate, MUW 1193, MY 93, Yukamate
IUPAC Name S-(2-phenylpropan-2-yl) piperidine-1-carbothioate
SMILES CC(C)(SC(=O)N1CCCCC1)c2ccccc2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H21NOS/c1-15(2,13-9-5-3-6-10-13)18-14(17)16-11-7-4-8-12-16/h3,5-6,9-10H,4,7-8,11-12H2,1-2H3
Formula Weight 263.1344
Chemical Name or Material Dimepiperate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.