missing translation for 'onlineSavingsMsg'
Learn More

Dimenhydrinate, TRC

Product Code. 30138357
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138357 5 g 5g
30010610 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138357 Supplier Toronto Research Chemicals Supplier No. TRCD4605205G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 523-87-5
Molecular Formula C17 H21 N O . C7 H7 Cl N4 O2
Molecular Weight (g/mol) 469.96
Synonym Dimenhydrinate, 1H-Purine-2,6-dione, 8-chloro-3,7-dihydro-1,3-dimethyl-, compd. with 2-(diphenylmethoxy)-N,N-dimethylethanamine (1:1), Theophylline, 8-chloro-, compd. with 2-(diphenylmethoxy)-N,N-dimethylethylamine (1:1) (7CI,8CI), Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, compd. with 8-chloro-3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione (1:1) (9CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, compd. with 8-chlorotheophylline (1:1) (8CI), 2-(Diphenylmethoxy)-N,N-dimethylethylamine 8-chlorotheophyllinate, Amosyt, Anautine, Andramine, Antemin, Aviomarin, Chloranautine, Diamarin, Dimate, Dimigal, Diphenhydramine 8-chlorotheophyllinate, Diphenhydrinate, Dommanate, Dramamin, Dramamine, Dramarin, Dramina, Dramocen, Dramyl, Dromyl, Emedyl, Emes, Epha, Faston, Gravinol, Gravinol (antiemetic), Gravol, Menhydrinate, NSC 117855, Neo-Navigan, Novamin, Novamine, Permital, Reidamine, Reise-Engletten, Removine, Supremal, Teodramin, Theohydramine, Travel-Gum, Travelin, Travelmin, Vomex A, Xamamina
IUPAC Name 2-benzhydryloxy-N,N-dimethylethanamine;8-chloro-1,3-dimethyl-7H-purine-2,6-dione
SMILES CN(C)CCOC(c1ccccc1)c2ccccc2.CN3C(=O)N(C)c4nc(Cl)[nH]c4C3=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C17H21NO.C7H7ClN4O2/c1-18(2)13-14-19-17(15-9-5-3-6-10-15)16-11-7-4-8-12-16;1-11-4-3(9-6(8)10-4)5(13)12(2)7(11)14/h3-12,17H,13-14H2,1-2H3;1-2H3,(H,9,10)
Formula Weight 469.1881
Chemical Name or Material Dimenhydrinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.