missing translation for 'onlineSavingsMsg'
Learn More

Dimenhydrinate, MedChemExpress

Product Code. 16478686
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478686 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16478686 Supplier Medchem Express Supplier No. HYB1215
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Dimenhydrinate is an anti-emetic and anti-histamine commonly available over-the-counter as a motion sickness remedy.

Dimenhydrinate is an anti-emetic and anti-histamine commonly available over-the-counter as a motion sickness remedy.

Specifications

Chemical Name or Material Dimenhydrinate
CAS 523-87-5
Color White
Purity Grade Notes Research
Molecular Formula C24H28ClN5O3
Quantity 100 mg
Solubility Information DMSO : ≥ 37 mg/mL (78.73 mM)
SMILES O=C(N1C)N(C)C2=C(NC(Cl)=N2)C1=O.CN(C)CCOC(C3=CC=CC=C3)C4=CC=CC=C4
Molecular Weight (g/mol) 469.96
Formula Weight 469.96
Percent Purity 99.69%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.