Learn More
Thermo Scientific™ DiluFlux Integral Stand for Broth Bags
Product Code. 17173784
Change view
Click to view available options
For Use With (Equipment):
DiluFlux Sample Dilution
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17173784 DiluFlux Sample Dilution Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17173784 Supplier Thermo Scientific™ Supplier No. DB4109A
  • ONLINE EXCLUSIVE PRICE
    296.00 EUR
    287.14 EUR / Pack of 1
    Save 8.86 EUR
    2% Off
 Restricted Item
Estimated Shipment: 08-12-2026
DiluFlux Stand for bagged media. Integrated to the unit, to hold one broth bag. Delivered with 4 screws + Torx screwdriver
Compatible with many prepared media formats, theeasy-to-clean, stainless steel DiluFlux Pro Dilutors are an ideal fit for any food testing laboratory.
Automated media dispensing and sample dilution
  • Compact and easy-to-use
  • Automated, rapid and accurate dispensing
  • Available with one or two pump heads for multiple media dispensing
  • Compatible with Dry-Bags and other media formats
TRUSTED_SUSTAINABILITY
Specifications
Product Type Bag Stand
For Use With (Equipment) DiluFlux Sample Dilution