missing translation for 'onlineSavingsMsg'
Learn More

Diloxanide furoate, MedChemExpress

Product Code. 16444557
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444557 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16444557 Supplier Medchem Express Supplier No. HYB1147
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Diloxanide furoate is the prodrug of Diloxanide. Diloxanide furoate is a potent and orally active anti-protozoal agent and can be used for the research of amebiasis, mild intestinal amebiasis or asymptomatic cyst carriers.

Diloxanide furoate is the prodrug of Diloxanide. Diloxanide furoate is a potent and orally active anti-protozoal agent and can be used for the research of amebiasis, mild intestinal amebiasis or asymptomatic cyst carriers.

Specifications

Chemical Name or Material Diloxanide furoate
Physical Form Powder
CAS 3736-81-0
Color Light Nude
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C14H11Cl2NO4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 50 mg/mL (152.37 mM) ∣Ethanol : 20 mg/mL (60.95 mM; Need ultrasonic)
SMILES O=C(C1=CC=CO1)OC2=CC=C(N(C(C(Cl)Cl)=O)C)C=C2
Molecular Weight (g/mol) 328.15
Formula Weight 328.15
Percent Purity 99.06%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.