Learn More
Diisopropylidene Acetone, TRC
Product Code. 30138768
Change view
Click to view available options
Quantity:
1 g
2.5 g
25 g
Unit Size:
1g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138768 25 g 25g
30133723 2.5 g 2.5g
30007059 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138768 Supplier Toronto Research Chemicals Supplier No. TRCD47341525G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 504-20-1
Molecular Formula C9H14O
Molecular Weight (g/mol) 138.21
Synonym Diisobutenyl Ketone, Diisopropylidene Acetone, Foron, NSC 38718, NSC 403517, Phorone, sym-Diisopropylidene Acetone, 2,6-Dimethyl-2,5-heptadien-4-one
SMILES O=C(/C=C(C)\C)/C=C(C)\C
Specifications
Quantity 25 g
InChI Formula InChI=1S/C9H14O/c1-7(2)5-9(10)6-8(3)4/h5-6H,1-4H3
Formula Weight 138.1045
Chemical Name or Material Diisopropylidene Acetone