missing translation for 'onlineSavingsMsg'
Learn More

Diisopropylamine, Sigma-Aldrich™

Product Code. 15876370
Change view
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876370 2.5 L 2.5 L
15839070 100 mL 100mL
15866370 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15876370 Supplier Sigma Aldrich Supplier No. 8036462500
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

Diisopropylamine (DIPA) is an aliphatic secondary amine commonly used as an organic base in chemical synthesis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-18-9

Specifications

Melting Point -70°C
Density 0.72 g/cm3 at 20°C
Boiling Point 83°C to 84°C/1013 hPa
Molecular Formula (CH3)2CHNHCH(CH3)2
MDL Number MFCD00008862
Quantity 2.5 L
Synonym Diisopropylamine, N,N-Diisopropylamine, Bis(1-methylethyl)amine, Diisopropylamine
Molecular Weight (g/mol) 101.19
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.