missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Ether, Supelco™

Product Code. 15886890
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15886890 10 L 10 L
15833970 1 L 1 L
15843970 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15886890 Supplier Supelco Supplier No. 1008676010
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™ ACS,Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-20-3

Specifications

Melting Point −85°C (lit.)
Density 0.725 g/mL at 25°C
Boiling Point 68°C to 69°C
Molecular Formula (CH3)2CHOCH(CH3)2
MDL Number MFCD00008880
Quantity 10 L
Synonym 2,2′-Oxybis[propane], 2,2-Propoxypropane, Isopropyl ether, Isopropyl ether
Molecular Weight (g/mol) 102.17
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.