missing translation for 'onlineSavingsMsg'
Learn More

Diisopropyl Ether, Sigma-Aldrich™

Product Code. 15888430
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15888430 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15888430 Supplier Sigma-Aldrich Supplier No. 8008662500
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stabilized with 2,6-di-tert-butyl-4-methylphenol (BHT for synthesis

Diisopropylether (DIPE) is secondary ether. Besides its high-octane number, it has low toxicity, low production cost, and good anti-knock properties. It is flammable, non-reactive, colorless, and slightly soluble in water but has a high solubility in organic compounds. Due to its ability to liquefy oil-based solutions, it is being used as a solvent in paints, waxes, dyes, and stain removal. DIPE serves as a solvent in various chemical processes, also used as an extractant and oxygenate in motor fuel.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-20-3

Specifications

Melting Point −85°C (lit.)
Density 0.725 g/mL at 25°C
Boiling Point 68°C to 69°C
Molecular Formula (CH3)2CHOCH(CH3)2
MDL Number MFCD00008880
Quantity 2.5 L
Synonym 2,2′-Oxybis[propane], 2,2-Propoxypropane, Isopropyl ether, Isopropyl ether
Molecular Weight (g/mol) 102.17
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.