Learn More
Diisobutylamine, 99%
Product Code. 11398276
Change view
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Unit Size:
1 L
100mL
500mL
Product Code. Quantity unitSize
11398276 1000 mL 1 L
11378276 100 mL 100mL
11388276 500 mL 500mL
3 options
Product Code. 11398276
Brand: Thermo Scientific Alfa Aesar L15495.AU
Request Bulk or Custom Format
CAS: 110-96-3 | C8H20N | 130.25 g/mol
Diisobutylamine was used to study the effect of achiral amine on hydrogenation of ethyl pyruvate over cinchonidine-Pt/Al2O3 catalyst system.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diisobutylamine was used to study the effect of achiral amine on hydrogenation of ethyl pyruvate over cinchonidine-Pt/Al2O3 catalyst system.
Solubility
Soluble in water 5 g/L (20°C).
Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 110-96-3
Molecular Formula C8H20N
Molecular Weight (g/mol) 130.25
MDL Number MFCD00008930
InChI Key OBYVIBDTOCAXSN-OCAPTIKFSA-O
Synonym diisobutylamine, 1-propanamine, 2-methyl-n-2-methylpropyl, bis 2-methylpropyl amine, amine, diisobutyl, bis beta-methylpropyl amine, di-isobutylamine, n,n-bis 2-methylpropyl amine, unii-t18y0a819s, ccris 6232, di-2-methylpropyl amine
PubChem CID 8085
IUPAC Name 2-methyl-N-(2-methylpropyl)propan-1-amine
SMILES CC[C@H](C)[NH2+][C@H](C)CC
Specifications
Melting Point -77°C
Density 0.744
Boiling Point 137°C to 139°C
Flash Point 30°C (86°F)
Refractive Index 1.409
Quantity 1000 mL
UN Number UN2361
Beilstein 1209251
Sensitivity Air Sensitive
Solubility Information Soluble in water 5g/L (20°C).
Formula Weight 129.25
Percent Purity 99%
Chemical Name or Material Diisobutylamine