missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl Phthalate, TRC

Product Code. 30179377
Change view
Click to view available options
Quantity:
1 g
50 g
Unit Size:
1g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179377 1 g 1g
30179253 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30179377 Supplier Toronto Research Chemicals Supplier No. TRCD4552101G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-69-5
Molecular Formula C16 H22 O4
Molecular Weight (g/mol) 278.34
Synonym 1,2-Benzenedicarboxylic acid, 1,2-bis(2-methylpropyl) ester, 1,2-Benzenedicarboxylic acid, bis(2-methylpropyl) ester (9CI), Phthalic acid, diisobutyl ester (6CI,7CI,8CI), 1,2-Benzenedicarboxylic acid diisobutyl ester, 2-Methylpropyl phthalate, Bis(2-methylpropyl) o-phthalate, Bis(2-methylpropyl) phthalate, Di(2-methylpropyl) phthalate, Di(isobutyl) 1,2-benzenedicarboxylate, Di-iso-Butyl phthalate, Diisobutyl phthalate, Hexaplas M/1B, Isobutyl phthalate, NSC 15316, Palatinol IC, Reomol DiBP
IUPAC Name bis(2-methylpropyl) benzene-1,2-dicarboxylate
SMILES CC(C)COC(=O)c1ccccc1C(=O)OCC(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H22O4/c1-11(2)9-19-15(17)13-7-5-6-8-14(13)16(18)20-10-12(3)4/h5-8,11-12H,9-10H2,1-4H3
Formula Weight 278.1518
Percent Purity >95%
Chemical Name or Material Phthalic acid, bis-iso-butyl ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.