missing translation for 'onlineSavingsMsg'
Learn More

Diisobutyl Phosphate, TRC

Product Code. 30138480
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138480 500 mg 500mg
30007454 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138480 Supplier Toronto Research Chemicals Supplier No. TRCD455480500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6303-30-6
Molecular Formula C8 H19 O4 P
Molecular Weight (g/mol) 210.21
Synonym Phosphoric Acid Bis(2-methylpropyl) Ester, Phosphoric Acid Diisobutyl Ester;
IUPAC Name diisobutyl hydrogen phosphate
SMILES CC(C)COP(=O)(O)OCC(C)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H19O4P/c1-7(2)5-11-13(9,10)12-6-8(3)4/h7-8H,5-6H2,1-4H3,(H,9,10)
Chemical Name or Material Diisobutyl Phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.