Learn More
Diiodochloromethane, TRC
Product Code. 30137295
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137295 100 mg 100mg
17975015 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137295 Supplier Toronto Research Chemicals Supplier No. TRCD466050100MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 638-73-3
Molecular Formula C H Cl I2
Molecular Weight (g/mol) 302.28
Synonym Chlorodiiodomethane, Diiodochloromethane, Methane, chlorodiiodo- (6CI,9CI)
IUPAC Name chloro(diiodo)methane
SMILES ClC(I)I
Specifications
Quantity 100 mg
InChI Formula InChI=1S/CHClI2/c2-1(3)4/h1H
Formula Weight 301.7856
Chemical Name or Material Diiodochloromethane