missing translation for 'onlineSavingsMsg'
Learn More

Dihydroresveratrol, TRC

Product Code. 30137702
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137702 50 mg 50mg
30135507 10 mg 10mg
30003791 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137702 Supplier Toronto Research Chemicals Supplier No. TRCD67896050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58436-28-5
Molecular Formula C14H14O3
Molecular Weight (g/mol) 230.26
Synonym 5-[2-(4-Hydroxyphenyl)ethyl]-1,3-benzenediol, 3,4',5-Trihydroxybibenzyl, 3,5,4'-Trihydroxybibenzyl, 5-(p-Hydroxyphenethyl)-resorcinol;
IUPAC Name 5-[2-(4-hydroxyphenyl)ethyl]benzene-1,3-diol
SMILES OC1=CC(CCC2=CC=C(O)C=C2)=CC(O)=C1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H14O3/c15-12-5-3-10(4-6-12)1-2-11-7-13(16)9-14(17)8-11/h3-9,15-17H,1-2H2
Formula Weight 230.0943
Chemical Name or Material Dihydroresveratrol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.