Learn More
Dihexadecyldimethylammonium Chloride, TRC
Artikelnummer. 30136232
Ansicht ändern
Click to view available options
Quantity:
100 mg
500 mg
1 g
Packungsgröße:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30136232 1 g 1g
30133681 500 mg 500mg
30040631 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30136232 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCD2685401G
 Restricted Item
Estimated Shipment: 27-08-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 1812-53-9
Molecular Formula C34 H72 N . Cl
Molecular Weight (g/mol) 530.4
Synonym Dimethyldipalmitylammonium Chloride, 1-Hexadecanaminium, N-hexadecyl-N,N-dimethyl-, chloride (1:1), 1-Hexadecanaminium, N-hexadecyl-N,N-dimethyl-, chloride (9CI), Ammonium, dihexadecyldimethyl-, chloride (8CI), Dihexadecyldimethylammonium chloride (6CI,7CI), Adogen 432-100, Adogen 432CG, Adogen 432PG, Aliquat 206, Ammonyx D 34, Dicetyldimethylammonium chloride, Dicetyldimonium chloride, Dimethyldicetylammonium chloride, Dimethyldihexadecylammonium chloride, Dimethyldipalmitylammonium chloride, Dipalmityldimethylammonium chloride, N,N-Dimethyl-N,N-dihexadecylammonium chloride, Varisoft 432PG, Varisoft 432PPG
IUPAC Name dihexadecyl(dimethyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCC
Spezifikation
Quantity 1 g
InChI Formula InChI=1S/C34H72N.ClH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35(3,4)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-34H2,1-4H3;1H/q+1;/p-1
Formula Weight 529.5353
Percent Purity >95%
Chemical Name or Material Dimethyldipalmitylammonium chloride
Grade HPLC