missing translation for 'onlineSavingsMsg'
Learn More

Dihexadecyl Phosphate, TRC

Product Code. 30370807
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370807 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370807 Supplier Toronto Research Chemicals Supplier No. TRCD4476005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dihexadecyl Phosphate
CAS 2197-63-9
Molecular Formula C32H67O4P
Quantity 5 g
Synonym 1-Hexadecanol, hydrogen phosphate (8CI,9CI), Bis(hexadecyl) phosphate, CS 20A, Crodafos CS 20A, DAP 60H, Di(hexadecyl) monohydrogen phosphate, Di-n-hexadecyl phosphate, Dicetyl hydrogen phosphate, Dicetyl phosphate, Dihexadecyl hydrogen phosphate, Dihexadecyl phosphate, NSC 12429, Phosphoric acid dihexadecyl ester, 1-Hexadecanol, 1,1'-(hydrogen phosphate)
InChI Formula InChI=1S/C32H67O4P/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-35-37(33,34)36-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-32H2,1-2H3,(H,33,34)
SMILES O=P(OCCCCCCCCCCCCCCCC)(O)OCCCCCCCCCCCCCCCC
IUPAC Name dihexadecyl hydrogen phosphate
Molecular Weight (g/mol) 546.85
Formula Weight 546.4777
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.