Learn More
Diglycerol (Mixture of Isomers), TRC
Product Code. 30136684
Change view
Click to view available options
Quantity:
5 g
50 g
100 g
Unit Size:
100g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136684 50 g 50g
30008176 5 g 5g
30006499 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136684 Supplier Toronto Research Chemicals Supplier No. TRCD44536050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 627-82-7
Molecular Formula C6H14O5
Molecular Weight (g/mol) 166.17
Synonym 3,3'-Oxydi-1,2-propanediol, 4-Oxaheptane-1,2,6,7-tetrol, Bis(2,3-dihydroxypropyl) Ether, NSC 8689
IUPAC Name 3,3'-oxybis(propane-1,2-diol)
SMILES OCC(O)COCC(O)CO
Specifications
Quantity 50 g
InChI Formula InChI=1S/C6H14O5/c7-1-5(9)3-11-4-6(10)2-8/h5-10H,1-4H2
Formula Weight 166.08
Chemical Name or Material Diglycerol