CAS RN 627-82-7
CAS RN 627-82-7
IUPAC Name:
3-(2,3-dihydroxypropoxy)propane-1,2-diol
Synonyms:
3-(2,3-dihydroxypropoxy)propane-1,2-diol
Molecular Weight (g/mol):
166.17
Molecular Formula:
C6H14O5
InChi Key:
GPLRAVKSCUXZTP-UHFFFAOYNA-N
SMILES:
OCC(O)COCC(O)CO
1
–
1
of
1
results
1
Diglycerol (Mixture of Isomers), TRC
CAS: 627-82-7 Molecular Formula: C6H14O5 Molecular Weight (g/mol): 166.17 Synonym: 3,3'-Oxydi-1,2-propanediol,4-Oxaheptane-1,2,6,7-tetrol,Bis(2,3-dihydroxypropyl) Ether,NSC 8689 IUPAC Name: 3,3'-oxybis(propane-1,2-diol) SMILES: OCC(O)COCC(O)CO