Learn More
TFA Dostmann Digital Thermo-Hygrometer
Product Code. 16436840
Change view
Click to view available options
Humidity Range:
20 to 95%
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Humidity Range Unit Size
16436840 20 to 95% Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16436840 Supplier TFA Dostmann Supplier No. 30.5027.02
  • ONLINE EXCLUSIVE PRICE
    11.56 EUR
    8.46 EUR / Pack of 1
    Save 3.10 EUR
    26% Off
Estimated Shipment: 17-12-2026
This modern ultra-flat glossy digital thermo-hygrometer is an ideal instrument for monitoring the indoor climate. You can easily keep an eye on temperature and humidity. If a smiley icon appears, the values are ideal. The highest and lowest values are stored and displayed at the push of a button.
At least two thirds of a person's lifetime are spent indoors. Whether we feel well and comfortably at home depends to a great extent on the quality of the indoor air. Rooms that are too humid favour mould growth, but overly dry air can be harmful to our health and pets, plants, wooden floors and antique furniture will also suffer. This modern ultra-flat glossy thermo-hygrometer is an ideal instrument for monitoring the indoor climate. You can easily keep an eye on temperature and humidity and regulate it by carefully heating and ventilating when the values are outside the comfort-level zone. This creates a pleasant and healthy living environment and even saves on heating costs. If a smiley icon appears, the values are ideal. The highest and lowest values are stored and displayed at the push of a button.
  • For a healthy indoor climate
  • Monitoring of indoor temperature and humidity
  • Max.-min. values
  • With Smiley as a comfort level indicator
  • Ultra-flat, high-gloss design
Specifications
Type Digital Thermohygrometer
Humidity Range 20 to 95%
Temperature Range (Metric) 0°C to 50°C