Learn More
TFA Dostmann™ Digital Indoor Thermo-Hygrometer
Product Code. 11882881
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
11882881 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11882881 Supplier TFA Dostmann™ Supplier No. 30.5015.02
Only null left
Add to basket
Add to basket
An excellent choice for the monitoring of indoor temperature and humidity.
  • Helps maintain a healthy indoor climate
  • Permanent indication of daily max. and min. values with auto reset
  • Additional max/min memory with manual reset
Specifications
Type Thermohygrometer
Humidity Range 20 to 99%
Temperature Range (Metric) -10°C to +70°C
Battery Type 1 x AAA 1.5 V
Display Type Digital
Length (Metric) 91 mm
Height (Metric) 134 mm
Weight (Metric) 100 g
Voltage 1.5 V
For Use With (Application) For inside and outside measurement
Includes Thermo-hygrometer, battery, instruction manual
Certifications/Compliance CE, EK-EL