Learn More
Diflunisal, Thermo Scientific Chemicals
Product Code. p-7077860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15567539 1 g 1g
15577539 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15567539 Supplier Thermo Scientific Acros Supplier No. 460890010
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 22494-42-4
Molecular Formula C13H8F2O3
Molecular Weight (g/mol) 250.20
MDL Number MFCD00057834
InChI Key HUPFGZXOMWLGNK-UHFFFAOYSA-N
Synonym diflunisal, dolobid, dolobis, fluniget, flovacil, fluodonil, adomal, flustar, 5-2,4-difluorophenyl salicylic acid, diflunisalum
PubChem CID 3059
ChEBI CHEBI:39669
IUPAC Name 5-(2,4-difluorophenyl)-2-hydroxybenzoic acid
SMILES OC(=O)C1=CC(=CC=C1O)C1=CC=C(F)C=C1F
Specifications
Melting Point 207.0°C to 209.0°C
Loss on Drying 0.5% max. (60°C, 4 H) (vacuum)
Infrared Spectrum Authentic
Linear Formula F2C6H3C6H3(OH)COOH
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol, dmso
Formula Weight 250.2
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Diflunisal
compliance-icons compliance-icons
Product Identifier
  • Diflunisal
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361-Suspected of damaging fertility or the unborn child.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.