Learn More
Masterflex™ Differential Pressure Flowmeters
Product Code. 15305184 Shop All Masterflex Products
Change view
Click to view available options
Flow Rate Range:
0.05 to 5 LPM
2 to 200 mL/min.
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Range Unit Size
15305184 0.05 to 5 LPM Each
12058588 2 to 200 mL/min. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15305184 Supplier Masterflex™ Supplier No. 3290869
  • ONLINE EXCLUSIVE PRICE
    2080.00 EUR
    1960.55 EUR / Pack of 1
    Save 119.45 EUR
    5% Off
Estimated Shipment: 11-12-2026
Specifications
Type Differential Pressure Flowmeter
Accuracy ±0.8% (Reading), ±0.2% (Full Scale)
Pressure Rating 145 psig
Connections 1/8 in. NPT (F)
Flow Rate Range 0.05 to 5 LPM
For Use With (Application) Versatility and high accuracy from a laminar-based mass flow meter
Power Supply Details 7 to 30 VDC at 30 mA
Repeatability ±0.5%
Temperature (Metric) 50°C