missing translation for 'onlineSavingsMsg'
Learn More

Diethyleneglycol Monolauryl Ether, TRC

Product Code. 30133211
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133211 500 mg 500mg
30009246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133211 Supplier Toronto Research Chemicals Supplier No. TRCD444385500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3055-93-4
Molecular Formula C16H34O3
Molecular Weight (g/mol) 274.44
Synonym 2-[2-(Dodecyloxy)ethoxy]ethanol, Bis(oxyethylene) Dodecyl Ether, C12EO2, Diethylene Glycol Dodecyl Ether, Diethylene Glycol Monododecyl Ether, Diethylene Glycol Monoether with Conol 20P, Diethyleneglycol Lauryl Ether, Dodecyl Diethylene Glycol, LA 2, LA 2 (Alcohol), LAE 2, Nikkol BL 2SY, Simulsol P 2
IUPAC Name 2-(2-dodecoxyethoxy)ethanol
SMILES CCCCCCCCCCCCOCCOCCO

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C16H34O3/c1-2-3-4-5-6-7-8-9-10-11-13-18-15-16-19-14-12-17/h17H,2-16H2,1H3
Formula Weight 274.2508
Chemical Name or Material Diethyleneglycol Monolauryl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.