CAS RN 3055-93-4
CAS RN 3055-93-4
IUPAC Name:
2-[2-(dodecyloxy)ethoxy]ethan-1-ol
Synonyms:
2-[2-(dodecyloxy)ethoxy]ethanol
Molecular Weight (g/mol):
274.45
Molecular Formula:
C16H34O3
InChi Key:
AZLWQVJVINEILY-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCOCCOCCO
1
–
1
of
1
results
1
Diethyleneglycol Monolauryl Ether, TRC
CAS: 3055-93-4 Molecular Formula: C16H34O3 Molecular Weight (g/mol): 274.44 Synonym: 2-[2-(Dodecyloxy)ethoxy]ethanol,Bis(oxyethylene) Dodecyl Ether,C12EO2,Diethylene Glycol Dodecyl Ether,Diethylene Glycol Monododecyl Ether,Diethylene Glycol Monoether with Conol 20P,Diethyleneglycol Lauryl Ether,Dodecyl Diethylene Glycol,LA 2,LA 2 (Alcohol),LAE 2,Nikkol BL 2SY,Simulsol P 2 IUPAC Name: 2-(2-dodecoxyethoxy)ethanol SMILES: CCCCCCCCCCCCOCCOCCO