missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Monobutyl Ether, Sigma-Aldrich™

Product Code. 15816350
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15816350 2.5 L 2.5 L
15806350 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15816350 Supplier Sigma Aldrich Supplier No. 8031292500
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-34-5

Specifications

Melting Point -68°C
Density 0.95 g/cm3 at 20°C
Boiling Point 226°C to 234°C/1013 hPa
Molecular Formula HOCH2CH2OCH2CH2O(CH2)3CH3
MDL Number MFCD00002881
Quantity 2.5 L
Synonym Diethylene glycol monobutyl ether, Butyl carbitol, 2-(2-Butoxyethoxy)ethanol, Butyl diglycol, Diethylene glycol monobutyl ether
Molecular Weight (g/mol) 162.23
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.