missing translation for 'onlineSavingsMsg'
Learn More

Diethylene Glycol Dimethacrylate, TRC

Product Code. 30377261
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377261 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30377261 Supplier Toronto Research Chemicals Supplier No. TRCD44435050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethylene Glycol Dimethacrylate
CAS 2358-84-1
Molecular Formula C12H18O5
Quantity 50 g
Synonym 2-Methyl-2-propenoic Acid 1,1'-(Oxydi-2,1-ethanediyl) Ester, Methacrylic Acid Oxydiethylene Ester, Diethylene Glycol Dimethacrylate, 2,2'-Oxybisethanol Dimethacrylate, 2,2'-Oxydiethyl Dimethacrylate, 2G, Blemmer PDE 100, DGM 2, Diethylene Glycol Bis(methacrylate), Light Ester 2EG, NK Ester 2G, Oxydiethylene Methacrylate, PDE 100, SR 231, SR 231E, TGM 2, Dipropyleneglycol Dimethylacrylate
InChI Formula InChI=1S/C12H18O5/c1-9(2)11(13)16-7-5-15-6-8-17-12(14)10(3)4/h1,3,5-8H2,2,4H3
SMILES CC(=C)C(=O)OCCOCCOC(=O)C(=C)C
IUPAC Name 2-[2-(2-methylprop-2-enoyloxy)ethoxy]ethyl 2-methylprop-2-enoate
Molecular Weight (g/mol) 242.27
Formula Weight 242.1154
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.