Learn More
Diethylene Glycol-d8, TRC
Specifications
Specifications
| Chemical Name or Material | Diethylene Glycol-d8 |
| CAS | 102867-56-1 |
| Molecular Formula | C4 2H8 H2 O3 |
| Quantity | 25 mg |
| Synonym | Diethylene Glycol D8, Ethan-1,1,2,2-d4-ol, 2,2'-oxybis-, 2,2'-Oxybis[ethan-1,1,2,2-d4-ol], 1,1,2,2-Tetradeuterio-2-(1,1,2,2-tetradeuterio-2-hydroxyethoxy)ethanol |
| InChI Formula | InChI=1S/C4H10O3/c5-1-3-7-4-2-6/h5-6H,1-4H2/i1D2,2D2,3D2,4D2 |
| SMILES | [2H]C([2H])(O)C([2H])([2H])OC([2H])([2H])C([2H])([2H])O |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-(1,1,2,2-tetradeuterio-2-hydroxyethoxy)ethanol |
| Molecular Weight (g/mol) | 114.17 |
| Formula Weight | 114.1132 |