missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine, Sigma-Aldrich™

Product Code. 15898890
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
50 L
Unit Size:
1 L
100mL
2.5 L
50 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15898890 500 mL 500mL
15886330 100 mL 100mL
15808900 1 L 1 L
15818900 2.5 L 2.5 L
15828900 50 L 50 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15898890 Supplier Sigma Aldrich Supplier No. 8030100500
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-89-7

Specifications

Melting Point -50°C
Density 0.71 g/cm3 at 20°C
Boiling Point 56°C/1013 mbar
Molecular Formula (C2H5)2NH
MDL Number MFCD00009032
Quantity 500 mL
Synonym Diethylamine, N-Ethylethanamine, Diethylamine
Molecular Weight (g/mol) 73.14
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.