missing translation for 'onlineSavingsMsg'
Learn More

Diethylamine Salicylate, TRC

Product Code. 30004751
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004751 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30004751 Supplier Toronto Research Chemicals Supplier No. TRCD4435151G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4419-92-5
Molecular Formula C7 H6 O3 . C4 H11 N
Quantity 1 g
Synonym Benzoic acid, 2-hydroxy-, compd. with N-ethylethanamine (1:1), Salicylic acid, Et2NH salt (6CI), Salicylic acid, compd. with Et2NH (7CI), Salicylic acid, compd. with diethylamine (1:1) (8CI), Diethylamine, compd. with salicylic acid (1:1) (8CI), Ethanamine, N-ethyl-, 2-hydroxybenzoate (9CI), Artrogota, Damsal, Diethylamine salicylate, Diethylammonium salicylate, Salicylate diethylamine, Salicylic acid diethylamine salt
InChI Formula InChI=1S/C7H6O3.C4H11N/c8-6-4-2-1-3-5(6)7(9)10;1-3-5-4-2/h1-4,8H,(H,9,10);5H,3-4H2,1-2H3
SMILES CCNCC.OC(=O)c1ccccc1O
IUPAC Name N-ethylethanamine;2-hydroxybenzoic acid
Molecular Weight (g/mol) 211.26
Formula Weight 211.1208
Chemical Name or Material Diethylammonium Salicylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.