missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diethyl Malonate, Sigma-Aldrich™
Specifications
Specifications
| Melting Point | -50°C |
| Density | 1.055 g/cm3 at 20°C |
| Boiling Point | 199°C/1013 hPa |
| Molecular Formula | CH2(COOC2H5)2 |
| MDL Number | MFCD00009195 |
| Quantity | 100 mL |
| Synonym | Diethyl malonate, Ethyl malonate, Malonic acid diethyl ester, Diethyl malonate |
| Molecular Weight (g/mol) | 160.17 |
| Grade | Research |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?