Learn More
Diethyl formamidomalonate, 97%, Thermo Scientific™
Product Code. 11384494
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384494 5 g 5g
11394494 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11384494 Supplier Thermo Scientific Alfa Aesar Supplier No. L01189.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6326-44-9
Molecular Formula C8H13NO5
Molecular Weight (g/mol) 203.194
MDL Number MFCD00003279
InChI Key PFLHGSJLYNJIOF-UHFFFAOYSA-N
Synonym diethyl formamidomalonate, diethyl 2-formamidomalonate, diethyl 2-formylaminomalonate, diethyl formaminomalonate, diethyl2-formylaminomalonate, 1,3-diethyl 2-formamidopropanedioate, diethyl 2-carbonylaminopropane-1,3-dioate, formamidomalonic acid, diethyl ester, malonic acid, formamido-, diethyl ester, diethylformaminomalonate
PubChem CID 80617
IUPAC Name diethyl 2-formamidopropanedioate
SMILES CCOC(=O)C(C(=O)OCC)NC=O
Specifikationer
Melting Point 50°C to 55°C
Boiling Point 173°C to 176°C (11 mmHg)
Quantity 5 g
Beilstein 1788903
Formula Weight 203.19
Percent Purity 97%
Chemical Name or Material Diethyl formamidomalonate
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.