Learn More
Diethyl Ether, Tested according to Ph.Eur., Honeywell™
Product Code. 15643510
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15643510 2.5 L 2.5 L
15633510 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15643510 Supplier Honeywell Riedel-de Haën Supplier No. 317012.5L
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 60-29-7
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00011646
InChI Key RTZKZFJDLAIYFH-UHFFFAOYSA-N
Synonym diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether
PubChem CID 3283
ChEBI CHEBI:35702
IUPAC Name ethoxyethane
SMILES CCOCC
Specifications
Color Colorless
Density 0.706g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.365
Flash Point −40°C (−40°F)
Ignition Point 320°F
Linear Formula (CH3CH2)2O
Vapor Pressure 2311 hPa (60°C)
UN Number 1155
Beilstein 1696894
Formula Weight 74.12g/mol
Chemical Name or Material Diethyl Ether, Tested according to Ph.Eur.