Learn More
Diethyl di-n-butylmalonate, 98+%, Thermo Scientific Chemicals
Product Code. 11302347
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11302347 5 g 5g
11312347 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11302347 Supplier Thermo Scientific Alfa Aesar Supplier No. L09902.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 596-75-8
Molecular Formula C15H28O4
Molecular Weight (g/mol) 272.385
MDL Number MFCD00026842
InChI Key WHKKUUPZLWUOIW-UHFFFAOYSA-N
Synonym diethyl dibutylmalonate, diethyl 2,2-dibutylmalonate, propanedioic acid, dibutyl-, diethyl ester, diethyl di-n-butylmalonate, diethyldibutylmalonate, malonic acid, dibutyl-, diethyl ester, propanedioic acid, 2,2-dibutyl-, 1,3-diethyl ester, 1,3-diethyl 2,2-dibutylpropanedioate, diethyl=dibutylmalonate, acmc-209mf2
PubChem CID 69000
IUPAC Name diethyl 2,2-dibutylpropanedioate
SMILES CCCCC(CCCC)(C(=O)OCC)C(=O)OCC
Specifications
Density 0.95
Boiling Point 150°C (12 mmHg)
Refractive Index 1.433
Quantity 5 g
Beilstein 518421
Formula Weight 272.39
Percent Purity ≥98%
Chemical Name or Material Diethyl di-n-butylmalonate