missing translation for 'onlineSavingsMsg'
Learn More

Diethyl cyanomethylphosphonate, 96%

Product Code. p-7080573
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449552 25 g 25g
11459552 100 g 100g
11469552 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449552 Supplier Thermo Scientific Alfa Aesar Supplier No. A10218.14
Add to basket

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

CAS: 2537-48-6 | C6H12NO3P | 177.14 g/mol

Diethyl cyanomethyl phosphonate is used as an intermediate in Horner-Emmons reaction for the synthesis of substituted nitriles and their amide and heterocyclic derivatives. It is known as a modified Wittig reagent used in the preparation of alpha, beta-unsaturated nitriles from ketones or aldehydes like 3-hydroxy-3-methylbutanal. It reacts with epoxides and nitrones to prepare cyano-substituted cyclopropanes and aziridines respectively. It is actively involved in the synthesis of alpha-arylated alkanenitriles via reaction with aryl iodides in presence of CuI.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl cyanomethyl phosphonate is used as an intermediate in Horner-Emmons reaction for the synthesis of substituted nitriles and their amide and heterocyclic derivatives. It is known as a modified Wittig reagent used in the preparation of alpha, beta-unsaturated nitriles from ketones or aldehydes like 3-hydroxy-3-methylbutanal. It reacts with epoxides and nitrones to prepare cyano-substituted cyclopropanes and aziridines respectively. It is actively involved in the synthesis of alpha-arylated alkanenitriles via reaction with aryl iodides in presence of CuI.

Solubility
Miscible with water, chloroform, terahydrofuran, ethyl acetate and methylene chloride.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 2537-48-6
Molecular Formula C6H12NO3P
Molecular Weight (g/mol) 177.14
MDL Number MFCD00001893
InChI Key KWMBADTWRIGGGG-UHFFFAOYSA-N
Synonym diethyl cyanomethylphosphonate, diethyl cyanomethyl phosphonate, diethylphosphono acetonitrile, cyanomethylphosphonic acid diethyl ester, diethylphosphonoacetonitrile, diethylcyanomethylphosphonate, diethyl cyanomethanephosphonate, phosphonic acid, cyanomethyl-, diethyl ester, cyanomethyl phosphonic acid diethyl ester
PubChem CID 75676
IUPAC Name 2-diethoxyphosphorylacetonitrile
SMILES CCOP(=O)(CC#N)OCC

Spezifikation

Density 1.094
Boiling Point 132°C to 135°C (1.2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.433
Quantity 25 g
UN Number UN3278
Beilstein 1772981
Solubility Information Miscible with water,chloroform,terahydrofuran,ethyl acetate and methylene chloride.
Formula Weight 177.14
Percent Purity ≥98%
Chemical Name or Material Diethyl cyanomethylphosphonate
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.