Learn More
Diethyl chlorophosphite, 97%, Thermo Scientific Chemicals
Product Code. 11332357
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11332357 5 g 5g
11342357 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11332357 Supplier Thermo Scientific Alfa Aesar Supplier No. L09919.06
 Restricted Item
In Stock
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Diethyl chlorophosphite acts as a versatile phosphorylating agent for ester and lactone enolates, amines and alkenols. It is used in the synthesis of organophosphates and are considered anticholinesterase agents through observation in studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethyl chlorophosphite acts as a versatile phosphorylating agent for ester and lactone enolates, amines and alkenols. It is used in the synthesis of organophosphates and are considered anticholinesterase agents through observation in studies.
Solubility
Soluble in DMSO and chloroform. Reacts with water.
Notes
Air & Moisture Sensitive. Store away from air and strong bases. Incompatible with water, bases, active metals and oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 589-57-1
Molecular Formula C4H10ClO2P
Molecular Weight (g/mol) 156.546
MDL Number MFCD00009074
InChI Key TXHWYSOQHNMOOU-UHFFFAOYSA-N
Synonym diethyl chlorophosphite, ethyl phosphorochloridite, diethyl phosphorochloridite, chlorodiethoxyphosphine, phosphorochloridous acid, diethyl ester, unii-f414s3v8k6, diethyl chlorophosphonite, chloro diethoxy phosphane, diethychlorophosphite, ethyl chlorophosphite
PubChem CID 68530
IUPAC Name chloro(diethoxy)phosphane
SMILES CCOP(OCC)Cl
Specifications
Density 1.082
Boiling Point 153°C to 155°C
Flash Point 25°C (77°F)
Odor Acrid
Refractive Index 1.4375
Quantity 5 g
UN Number UN2920
Beilstein 1098392
Sensitivity Air and moisture sensitive
Merck Index 14,3841
Solubility Information Soluble in DMSO and chloroform. Reacts with water.
Formula Weight 156.55
Percent Purity 97%
Chemical Name or Material Diethyl chlorophosphite
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Diethyl chlorophosphite
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Diethyl chlorophosphite