Learn More
Diethyl (Boc-amino)malonate, TRC
Specifications
Specifications
| Chemical Name or Material | Diethyl (Boc-amino)malonate |
| CAS | 102831-44-7 |
| Molecular Formula | C12 H21 N O6 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C12H21NO6/c1-6-17-9(14)8(10(15)18-7-2)13-11(16)19-12(3,4)5/h8H,6-7H2,1-5H3,(H,13,16) |
| SMILES | CCOC(=O)C(NC(=O)OC(C)(C)C)C(=O)OCC |
| IUPAC Name | diethyl 2-[(2-methylpropan-2-yl)oxycarbonylamino]propanedioate |
| Molecular Weight (g/mol) | 275.3 |
| Formula Weight | 275.1369 |