Learn More
Diethyl 2-Formamidopropanedioate, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Diethyl 2-Formamidopropanedioate |
| CAS | 6326-44-9 |
| Molecular Formula | C8H13NO5 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C8H13NO5/c1-3-13-7(11)6(9-5-10)8(12)14-4-2/h5-6H,3-4H2,1-2H3,(H,9,10) |
| SMILES | CCOC(=O)C(NC=O)C(=O)OCC |
| IUPAC Name | diethyl 2-formamidopropanedioate |
| Molecular Weight (g/mol) | 203.19 |
| Formula Weight | 203.0794 |