Learn More
Diethyl (2-bromoethyl)phosphonate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 5324-30-1 |
| Molecular Formula | C6H14BrO3P |
| MDL Number | MFCD00000239 |
| Quantity | 1 g |
| SMILES | BrCCP(OCC)(OCC)=O |
| Molecular Weight (g/mol) | 245.05 |
| Percent Purity | 98% |
| Chemical Name or Material | Diethyl (2-bromoethyl)phosphonate |