missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate, 97%

Product Code. 11489317
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11489317 25 g 25g
11479317 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489317 Supplier Alfa Aesar™ Supplier No. B21374.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2436-79-5
Molecular Formula C12H17NO4
Molecular Weight (g/mol) 239.271
MDL Number MFCD00005218
InChI Key XSBSXJAYEPDGSF-UHFFFAOYSA-N
PubChem CID 75526
IUPAC Name diethyl 3,5-dimethyl-1H-pyrrole-2,4-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C1C)C(=O)OCC)C

Specifications

Melting Point 135°C to 138°C
Quantity 25 g
Synonym diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate, 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylic acid diethyl ester, knorr's pyrrole, 2,4-diethyl 3,5-dimethyl-1h-pyrrole-2,4-dicarboxylate, 1h-pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, 2,4-dimethyl-3,5-dicarbethoxypyrrole, diethyl 2,4-dimethyl-1h-pyrrole-3,5-dicarboxylate, diethyl2,4-dimethylpyrrole-3,5-dicarboxylate, pyrrole-2,4-dicarboxylic acid, 3,5-dimethyl-, diethyl ester, diethyl 2 ,4-dimethylpyrrole-3,5-dicarboxylate
Formula Weight 239.27
Percent Purity 97%
Chemical Name or Material Diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.