Learn More
Diethoxymethylvinylsilane, 97%
Product Code. 11359055
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359055 25 g 25g
11349055 5 g 5g
11369055 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11359055 Supplier Thermo Scientific Alfa Aesar Supplier No. L14224.14
Request Bulk or Custom Format
CAS: 5507-44-8 | C7H16O2Si | 160.288 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5507-44-8
Molecular Formula C7H16O2Si
Molecular Weight (g/mol) 160.288
MDL Number MFCD00009067
InChI Key MBGQQKKTDDNCSG-UHFFFAOYSA-N
Synonym diethoxy methyl vinyl silane, methylvinyldiethoxysilane, silane, ethenyldiethoxymethyl, vinylmethyldiethoxysilane, diethoxy methyl vinylsilane, silane, diethoxymethylvinyl, vinyldiethoxymethylsilane, vinylmethyldiathoxysilan, diethoxymethylvinyl silane, diethoxy-methyl-vinyl-silane
PubChem CID 79646
IUPAC Name ethenyl-diethoxy-methylsilane
SMILES CCO[Si](C)(C=C)OCC
Specifications
Boiling Point 133°C to 134°C
Quantity 25 g
UN Number UN1993
Beilstein 970124
Flash Point 17°C (62°F)
Formula Weight 160.29
Percent Purity 97%
Refractive Index 1.4
Sensitivity Moisture sensitive
Density 0.858
Chemical Name or Material Diethoxymethylvinylsilane