accessibility menu, dialog, popup

UserName

CAS RN 5507-44-8

CH3OSiCH3OH3CH2C

CAS RN 5507-44-8

IUPAC Name: ethenyldiethoxymethylsilane
Synonyms: silane, ethenyldiethoxymethyl-
Molecular Weight (g/mol): 160.29
Molecular Formula: C7H16O2Si
InChi Key: MBGQQKKTDDNCSG-UHFFFAOYSA-N
SMILES: CCO[Si](C)(OCC)C=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results