Learn More
Diethanolamine, Supelco™
Product Code. 15815860
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15815860 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15815860 Supplier Supelco Supplier No. 1162051000
 Restricted Item
Estimated Shipment: 14-12-2026
For analysis EMSURE™
EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
TRUSTED_SUSTAINABILITY
Specifications
Melting Point 28°C
CAS 111-42-2
Density 1.09 g/cm3 at 30°C (Liquid)
Quantity 1 L
Molecular Formula NH(CH2CH2OH)2
MDL Number MFCD00002843
Synonym Diethanolamine, β,β′-Dihydroxydiethylamine, Bis-(β-hydroxyethyl)-amine, Diethanolamine
Boiling Point 269°C to 270°C/1013 hPa
Molecular Weight (g/mol) 105.14
Grade Research
Chemical Name or Material Diethanolamine