Learn More
Diethanolamine, Sigma-Aldrich™
Product Code. 15866340
Change view
Click to view available options
Quantity:
1 L
5 L
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15866340 1 L 1 L
15838930 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15866340 Supplier Sigma Aldrich Supplier No. 8031161000
 Restricted Item
Estimated Shipment: 18-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 111-42-2
Specifications
Melting Point 28°C
Density 1.09 g/cm3 at 30°C (liquid)
Boiling Point 269°C to 270°C/1013 hPa
Molecular Formula NH(CH2CH2OH)2
MDL Number MFCD00002843
Quantity 1 L
Synonym Diethanolamine, β,β′-Dihydroxydiethylamine, Bis-(β-hydroxyethyl)-amine, Diethanolamine
Molecular Weight (g/mol) 105.14
Grade Research