Learn More
Dienogest, MedChemExpress
Product Code. 16468338
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468338 10 mg 10mg
16424227 50 mg 50mg
16418086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16468338 Supplier Medchem Express Supplier No. HYB008410mg
MedChemExpress Dienogest(STS-557) is a specific progesterone receptor agonist with potent oral endometrial activity and is used in the treatment of endometriosis.
Dienogest(STS-557) is a specific progesterone receptor agonist with potent oral endometrial activity and is used in the treatment of endometriosis.
Specifications
Chemical Name or Material Dienogest
Physical Form Solid
CAS 65928-58-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H25NO2
For Use With (Application) Cancer-programmed cell death
Synonym STS 557
Solubility Information DMSO : 25 mg/mL (80.28 mM; Need ultrasonic)
SMILES C[C@@]12[C@](O)(CC#N)CC[C@@]1([H])[C@]3([H])CCC4=CC(CCC4=C3CC2)=O
Molecular Weight (g/mol) 311.42
Formula Weight 311.42
Percent Purity 99.7%
Grade Research